Search
Search

gsx2| Gene Symbol: gsx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025512.2 | NP_001020683.1 | 1 - 2 | 483 | 64 | |
| GenBank mRNA | BC115063.1 | - | 1 - 2 | 483 | 64 | |
| BC164330.1 | - | 1 - 2 | 420 | 64 | ||
Set Membership: |
|