Search Thermo Fisher Scientific
Gene Symbol: tspan3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025538.2 | NP_001020709.1 | 4 - 5 | 534 | 65 | |
GenBank mRNA | BC097114.1 | - | 4 - 5 | 531 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |