Search
Search

furinb| Gene Symbol: furinb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045109.2 | NP_001038574.2 | 4 - 5 | 924 | 84 | |
| GenBank mRNA | BC163243.1 | - | 4 - 5 | 634 | 84 | |
| DQ499477.1 | - | 4 - 5 | 884 | 84 | ||
Set Membership: |
|