Search
Search

foxe3| Gene Symbol: foxe3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079682.2 | NP_001073150.2 | 1 | 1247 | 90 | |
| GenBank mRNA | BC163348.1 | - | 1 | 1210 | 90 | |
| BC163364.1 | - | 1 | 1210 | 90 | ||
| DQ457020.1 | - | 1 | 1247 | 90 | ||
Set Membership: |
|