Search
Search

nrxn2a| Gene Symbol: nrxn2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079970.1 | NP_001073439.1 | 23 - 24 | 4918 | 76 | |
| XM_009295476.3 | XP_009293751.1 | 23 - 24 | 5119 | 76 | ||
| XM_009295477.3 | XP_009293752.1 | 22 - 23 | 5095 | 76 | ||
| XM_009295479.3 | XP_009293754.1 | 6 - 7 | 2382 | 76 | ||
| XM_009295480.3 | XP_009293755.1 | 5 - 6 | 2292 | 76 | ||
| XM_021468849.1 | XP_021324524.1 | 23 - 24 | 5143 | 76 | ||
| XM_021468850.1 | XP_021324525.1 | 23 - 24 | 4909 | 76 | ||
| XM_021468851.1 | XP_021324526.1 | 22 - 23 | 5116 | 76 | ||
| XM_021468852.1 | XP_021324527.1 | 21 - 22 | 4196 | 76 | ||
| XM_021468853.1 | XP_021324528.1 | 21 - 22 | 5074 | 76 | ||
| XM_021468854.1 | XP_021324529.1 | 22 - 23 | 5054 | 76 | ||
| GenBank mRNA | DQ641428.1 | - | 23 - 24 | 4918 | 76 | |
| DQ641429.1 | - | 6 - 7 | 2321 | 76 | ||
Set Membership: |
|