Search
Search

git2b| Gene Symbol: git2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080188.2 | NP_001073657.2 | 16 - 17 | 1783 | 146 | |
| XM_005155407.4 | XP_005155464.1 | 17 - 18 | 1875 | 146 | ||
| GenBank mRNA | BC129227.1 | - | 16 - 17 | 1782 | 146 | |
Set Membership: |
|