Search
Search

cables1| Gene Symbol: cables1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105665.1 | NP_001099135.1 | 8 - 9 | 1293 | 64 | |
| XM_005163651.4 | XP_005163708.1 | 7 - 8 | 1740 | 64 | ||
| XM_021478424.1 | XP_021334099.1 | 7 - 8 | 1743 | 64 | ||
| GenBank mRNA | EF105292.1 | - | 8 - 9 | 1293 | 64 | |
Set Membership: |
|