Search
Search

nipsnap1| Gene Symbol: nipsnap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131033.2 | NP_571108.2 | 9 - 10 | 1067 | 97 | |
| GenBank mRNA | AJ249796.1 | - | 9 - 10 | 767 | 97 | |
| BC122249.1 | - | 9 - 10 | 1067 | 97 | ||
| EH486911.1 | - | 1 - 2 | 430 | 97 | ||
| EH513303.1 | - | 9 - 10 | 826 | 97 | ||
Set Membership: |
|