Search
Search

ctnna1| Gene Symbol: ctnna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131456.1 | NP_571531.1 | 12 - 13 | 2036 | 69 | |
| GenBank mRNA | AF099737.1 | - | 12 - 13 | 2036 | 69 | |
| BC066669.1 | - | 12 - 13 | 1963 | 69 | ||
| BC155815.1 | - | 12 - 13 | 1994 | 69 | ||
Set Membership: |
|