Search
Search

hoxc13a| Gene Symbol: hoxc13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131543.1 | NP_571618.1 | 1 - 2 | 699 | 79 | |
| GenBank mRNA | AY452737.1 | - | 1 - 2 | 699 | 79 | |
| BC134023.1 | - | 1 - 2 | 694 | 79 | ||
| DQ060560.1 | - | 1 - 2 | 322 | 79 | ||
Set Membership: |
|