Search
Search

cry1b| Gene Symbol: cry1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131790.4 | NP_571865.4 | 10 - 11 | 1841 | 82 | |
| GenBank mRNA | AB042249.1 | - | 9 - 10 | 1496 | 82 | |
| BC044558.1 | - | 10 - 11 | 1841 | 82 | ||
Set Membership: |
|