Search
Search

lamc1| Gene Symbol: lamc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173277.1 | NP_775384.1 | 6 - 7 | 1648 | 61 | |
| GenBank mRNA | AF468048.1 | - | 6 - 7 | 1648 | 61 | |
| BC068426.1 | - | 6 - 7 | 812 | 61 | ||
Set Membership: |
|