Search
Search

prickle1a| Gene Symbol: prickle1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183342.2 | NP_899185.2 | 6 - 7 | 983 | 90 | |
| XM_005174210.4 | XP_005174267.1 | 6 - 7 | 1072 | 90 | ||
| GenBank mRNA | AY278986.1 | - | 5 - 6 | 802 | 90 | |
| AY286492.1 | - | 6 - 7 | 983 | 90 | ||
| BC162516.1 | - | 5 - 6 | 829 | 90 | ||
Set Membership: |
|