Search
Search

eif5| Gene Symbol: eif5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199591.2 | NP_955885.1 | 9 - 10 | 1475 | 76 | |
| XM_005160454.3 | XP_005160511.1 | 10 - 11 | 1599 | 76 | ||
| GenBank mRNA | AY577010.1 | - | 10 - 11 | 1542 | 76 | |
| BC049502.1 | - | 8 - 9 | 1114 | 76 | ||
| BC066614.1 | - | 9 - 10 | 1475 | 76 | ||
Set Membership: |
|