Search
Search

aldh9a1b| Gene Symbol: aldh9a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201508.1 | NP_958916.1 | 3 - 4 | 373 | 81 | |
| XM_005163231.4 | XP_005163288.1 | 3 - 4 | 81 | |||
| XM_009297055.3 | XP_009295330.1 | 3 - 4 | 81 | |||
| GenBank mRNA | BC047176.1 | - | 3 - 4 | 373 | 81 | |
| BC165758.1 | - | 2 - 3 | 361 | 81 | ||
Set Membership: |
|