Search
Search

pvalb6| Gene Symbol: pvalb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205573.1 | NP_991136.1 | 2 - 3 | 279 | 105 | |
| XM_021469917.1 | XP_021325592.1 | 2 - 3 | 111 | 105 | ||
| GenBank mRNA | AF467913.1 | - | 2 - 3 | 279 | 105 | |
| BC081523.1 | - | 2 - 3 | 297 | 105 | ||
| BC164914.1 | - | 1 - 2 | 97 | 105 | ||
Set Membership: |
|