Search
Search

elf2a| Gene Symbol: elf2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004116.1 | NP_001004116.1 | 6 - 7 | 897 | 103 | |
| XM_005169610.4 | XP_005169667.1 | 6 - 7 | 935 | 103 | ||
| XM_009291253.3 | XP_009289528.1 | 4 - 5 | 360 | 103 | ||
| GenBank mRNA | BC080260.1 | - | 6 - 7 | 897 | 103 | |
Set Membership: |
|