Search
Search

mtdha| Gene Symbol: mtdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007135.2 | NP_001007136.2 | 5 - 6 | 947 | 77 | |
| XM_005159603.3 | XP_005159660.1 | 5 - 6 | 1023 | 77 | ||
| XM_005159604.3 | XP_005159661.1 | 5 - 6 | 1024 | 77 | ||
| XM_017352195.2 | XP_017207684.1 | 5 - 6 | 1027 | 77 | ||
| GenBank mRNA | BC068430.1 | - | 5 - 6 | 947 | 77 | |
Set Membership: |
|