Search
Search

mcamb| Gene Symbol: mcamb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012367.2 | NP_001012367.2 | 5 - 6 | 674 | 64 | |
| XM_005157570.2 | XP_005157627.1 | 5 - 6 | 697 | 64 | ||
| GenBank mRNA | AY497061.1 | - | 5 - 6 | 653 | 64 | |
| AY746594.1 | - | 5 - 6 | 674 | 64 | ||
| BC162589.1 | - | 5 - 6 | 666 | 64 | ||
Set Membership: |
|