Search
Search

cyp2x6| Gene Symbol: cyp2x6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013565.2 | NP_001013583.1 | 3 - 4 | 515 | 145 | |
| GenBank mRNA | BC078308.1 | - | 3 - 4 | 520 | 145 | |
| BC090406.1 | - | 3 - 4 | 519 | 145 | ||
Set Membership: |
|