Search
Search

kdr| Gene Symbol: kdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024653.2 | NP_001019824.2 | 6 - 7 | 1036 | 61 | |
| GenBank mRNA | AY523999.1 | - | 6 - 7 | 1007 | 61 | |
| DQ026829.1 | - | 6 - 7 | 1036 | 61 | ||
Set Membership: |
|