Search
Search

lmx1ba| Gene Symbol: lmx1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025168.2 | NP_001020339.1 | 3 - 4 | 767 | 86 | |
| NM_001362592.1 | NP_001349521.1 | 3 - 4 | 773 | 86 | ||
| GenBank mRNA | AY551078.1 | - | 3 - 4 | 478 | 86 | |
| AY894990.1 | - | 3 - 4 | 783 | 86 | ||
| BC163321.1 | - | 3 - 4 | 651 | 86 | ||
| BC163333.1 | - | 3 - 4 | 651 | 86 | ||
Set Membership: |
|