Search
Search

tprb| Gene Symbol: tprb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030123.1 | NP_001025294.1 | 35 - 36 | 5200 | 77 | |
| XM_009294751.3 | XP_009293026.1 | 35 - 36 | 5221 | 77 | ||
Set Membership: |
|