Search
Search

thbs3b| Gene Symbol: thbs3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045214.1 | NP_001038679.1 | 21 - 22 | 2586 | 70 | |
| XM_005170062.4 | XP_005170119.1 | 21 - 22 | 2997 | 70 | ||
| GenBank mRNA | BC163250.1 | - | 20 - 21 | 2544 | 70 | |
Set Membership: |
|