Search
Search

loxl5b| Gene Symbol: loxl5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102574.1 | NP_001096044.1 | 3 - 4 | 963 | 70 | |
| XM_017357369.2 | XP_017212858.1 | 3 - 4 | 1115 | 70 | ||
| XM_017357372.2 | XP_017212861.1 | 4 - 5 | 1580 | 70 | ||
| GenBank mRNA | EF030486.1 | - | 3 - 4 | 963 | 70 | |
Set Membership: |
|