Search
Search

elf1| Gene Symbol: elf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131159.2 | NP_571234.2 | 2 - 3 | 421 | 77 | |
| XM_017358977.2 | XP_017214466.1 | 2 - 3 | 738 | 77 | ||
| XM_017358978.2 | XP_017214467.1 | 2 - 3 | 533 | 77 | ||
| XM_017358979.2 | XP_017214468.1 | 2 - 3 | 716 | 77 | ||
| XM_017358980.2 | XP_017214469.1 | 2 - 3 | 738 | 77 | ||
| GenBank mRNA | U84615.1 | - | 2 - 3 | 427 | 77 | |
Set Membership: |
|