Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: calm3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182967.1 | NP_892012.1 | 5 - 6 | 538 | 102 | |
GenBank mRNA | BC045298.1 | - | 5 - 6 | 538 | 102 | |
BC071404.1 | - | 5 - 6 | 564 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |