Search
Search

slc1a2b| Gene Symbol: slc1a2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199979.2 | NP_956273.2 | 9 - 10 | 1650 | 81 | |
| XM_009298220.3 | XP_009296495.1 | 9 - 10 | 1435 | 81 | ||
| XM_009298221.3 | XP_009296496.1 | 9 - 10 | 1414 | 81 | ||
| XM_009298222.3 | XP_009296497.1 | 9 - 10 | 1601 | 81 | ||
| GenBank mRNA | BC056751.1 | - | 9 - 10 | 1575 | 81 | |
| BC155822.1 | - | 9 - 10 | 1600 | 81 | ||
| HM138691.1 | - | 8 - 9 | 1392 | 81 | ||
Set Membership: |
|