Search
Search

phactr4b| Gene Symbol: phactr4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200670.1 | NP_956964.1 | 11 - 12 | 2184 | 64 | |
| XM_005158332.4 | XP_005158389.1 | 10 - 11 | 2569 | 64 | ||
| XM_005158333.4 | XP_005158390.1 | 10 - 11 | 2797 | 64 | ||
| XM_005158334.4 | XP_005158391.1 | 12 - 13 | 2563 | 64 | ||
| XM_005158335.4 | XP_005158392.1 | 9 - 10 | 2435 | 64 | ||
| XM_009292432.3 | XP_009290707.1 | 12 - 13 | 3063 | 64 | ||
| XM_009292434.3 | XP_009290709.1 | 11 - 12 | 2431 | 64 | ||
| XM_021466650.1 | XP_021322325.1 | 12 - 13 | 2511 | 64 | ||
| GenBank mRNA | BC058052.1 | - | 11 - 12 | 2184 | 64 | |
Set Membership: |
|