Search
Search

abcf2a| Gene Symbol: abcf2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201315.2 | NP_958472.2 | 4 - 5 | 697 | 66 | |
| GenBank mRNA | BC047181.1 | - | 4 - 5 | 672 | 66 | |
| BC066505.1 | - | 4 - 5 | 601 | 66 | ||
| BC165771.1 | - | 3 - 4 | 576 | 66 | ||
| EH577748.1 | - | 4 - 5 | 452 | 66 | ||
Set Membership: |
|