Search
Search

rpp14| Gene Symbol: rpp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020692.2 | NP_001018528.1 | 5 - 6 | 437 | 108 | |
| NR_144413.1 | 5 - 6 | 450 | 108 | |||
| XM_005173956.4 | XP_005174013.1 | 5 - 6 | 422 | 108 | ||
| GenBank mRNA | BC095792.1 | - | 5 - 6 | 407 | 108 | |
| BC165532.1 | - | 4 - 5 | 344 | 108 | ||
| EH542438.1 | - | 5 - 6 | 458 | 108 | ||
| EH595656.1 | - | 5 - 6 | 471 | 108 | ||
Set Membership: |
|