Search
Search

slc43a3b| Gene Symbol: slc43a3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039922.1 | NP_001035011.1 | 8 - 9 | 1081 | 99 | |
| XM_009291002.3 | XP_009289277.1 | 9 - 10 | 1218 | 99 | ||
| GenBank mRNA | BC114300.1 | - | 8 - 9 | 1081 | 99 | |
Set Membership: |
|