Search
Search

rcor1| Gene Symbol: rcor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080041.1 | NP_001073510.1 | 8 - 9 | 888 | 82 | |
| GenBank mRNA | BC129322.1 | - | 8 - 9 | 888 | 82 | |
| BC165590.1 | - | 8 - 9 | 800 | 82 | ||
Set Membership: |
|