Search
Search

adat2| Gene Symbol: adat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083097.1 | NP_001076566.1 | 4 - 5 | 540 | 81 | |
| XM_005160618.4 | XP_005160675.1 | 4 - 5 | 629 | 81 | ||
| GenBank mRNA | BC135109.1 | - | 4 - 5 | 538 | 81 | |
| BC160619.1 | - | 4 - 5 | 541 | 81 | ||
| BC163783.1 | - | 4 - 5 | 513 | 81 | ||
| BC163804.1 | - | 4 - 5 | 513 | 81 | ||
Set Membership: |
|