Search
Search

pax6b| Gene Symbol: pax6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131641.1 | NP_571716.1 | 11 - 12 | 1491 | 85 | |
| XM_005168819.4 | XP_005168876.1 | 12 - 13 | 1557 | 85 | ||
| XM_005168823.4 | XP_005168880.1 | 9 - 10 | 1454 | 85 | ||
| XM_017356906.2 | XP_017212395.1 | 11 - 12 | 1499 | 85 | ||
| XM_021477600.1 | XP_021333275.1 | 12 - 13 | 1499 | 85 | ||
| XM_021477601.1 | XP_021333276.1 | 13 - 14 | 1628 | 85 | ||
| XM_021477602.1 | XP_021333277.1 | 12 - 13 | 1454 | 85 | ||
| XM_021477603.1 | XP_021333278.1 | 12 - 13 | 1529 | 85 | ||
| GenBank mRNA | AF061252.1 | - | 11 - 12 | 1491 | 85 | |
| BC076068.1 | - | 11 - 12 | 1353 | 85 | ||
Set Membership: |
|