Search
Search

sf3b4| Gene Symbol: sf3b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153661.4 | NP_705947.3 | 4 - 5 | 1064 | 152 | |
| GenBank mRNA | AY099515.2 | - | 4 - 5 | 1055 | 152 | |
| BC056532.1 | - | 4 - 5 | 1050 | 152 | ||
| BC067655.1 | - | 4 - 5 | 1054 | 152 | ||
| CT611775.2 | - | 1 - 2 | 332 | 152 | ||
Set Membership: |
|