Search
Search

rps21| Gene Symbol: rps21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201191.1 | NP_957485.1 | 4 - 5 | 255 | 71 | |
| GenBank mRNA | BC049056.1 | - | 4 - 5 | 255 | 71 | |
| BC071475.1 | - | 4 - 5 | 255 | 71 | ||
Set Membership: |
|