Search
Search

cyp2p10| Gene Symbol: cyp2p10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201511.1 | NP_958919.1 | 3 - 4 | 421 | 92 | |
| XM_005160559.4 | XP_005160616.1 | 2 - 3 | 388 | 92 | ||
| GenBank mRNA | BC049521.1 | - | 3 - 4 | 421 | 92 | |
Set Membership: |
|