Search
Search

hnrnpdl| Gene Symbol: hnrnpdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328494.1 | NP_001315423.1 | 6 - 7 | 962 | 109 | |
| NR_137312.1 | 6 - 7 | 962 | 109 | |||
| XM_021479148.1 | XP_021334823.1 | 6 - 7 | 967 | 109 | ||
| XM_021479149.1 | XP_021334824.1 | 6 - 7 | 967 | 109 | ||
| GenBank mRNA | BC055525.1 | - | 6 - 7 | 962 | 109 | |
| EH562663.1 | - | 2 - 3 | 382 | 109 | ||
| EH585301.1 | - | 3 - 4 | 407 | 109 | ||
Set Membership: |
|