Search
Search

msrb3| Gene Symbol: msrb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002094.2 | NP_001002094.2 | 2 - 3 | 287 | 143 | |
| XM_021474986.1 | XP_021330661.1 | 3 - 4 | 452 | 143 | ||
| GenBank mRNA | BC071530.1 | - | 2 - 3 | 244 | 143 | |
Set Membership: |
|