Search
Search

tspan18b| Gene Symbol: tspan18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002439.1 | NP_001002439.1 | 8 - 9 | 936 | 67 | |
| XM_005159172.4 | XP_005159229.1 | 8 - 9 | 1215 | 67 | ||
| XM_005159173.4 | XP_005159230.1 | 8 - 9 | 825 | 67 | ||
| XM_017351914.1 | XP_017207403.1 | 7 - 8 | 1050 | 67 | ||
| GenBank mRNA | BC076124.1 | - | 8 - 9 | 936 | 67 | |
Set Membership: |
|