Search
Search

trpa1b| Gene Symbol: trpa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007066.2 | NP_001007067.1 | 24 - 25 | 2970 | 73 | |
| XM_009297331.3 | XP_009295606.1 | 24 - 25 | 3296 | 73 | ||
| XM_021470156.1 | XP_021325831.1 | 24 - 25 | 3313 | 73 | ||
| GenBank mRNA | AY677197.1 | - | 24 - 25 | 2975 | 73 | |
| EU826642.1 | - | 23 - 24 | 2848 | 73 | ||
Set Membership: |
|