Search
Search

blvra| Gene Symbol: blvra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076601.1 | NP_001070069.1 | 6 - 7 | 731 | 63 | |
| GenBank mRNA | BC124225.1 | - | 6 - 7 | 731 | 63 | |
| BC163949.1 | - | 5 - 6 | 649 | 63 | ||
Set Membership: |
|