Search
Search

ptprfb| Gene Symbol: ptprfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083576.1 | NP_001077045.1 | 12 - 13 | 2689 | 60 | |
| XM_009298457.3 | XP_009296732.1 | 15 - 16 | 2779 | 60 | ||
| XM_009298458.3 | XP_009296733.1 | 15 - 16 | 2567 | 60 | ||
| XM_009298459.3 | XP_009296734.1 | 15 - 16 | 2779 | 60 | ||
| XM_009298460.3 | XP_009296735.1 | 14 - 15 | 2767 | 60 | ||
| XM_009298461.3 | XP_009296736.1 | 14 - 15 | 2761 | 60 | ||
| XM_009298462.3 | XP_009296737.1 | 13 - 14 | 2755 | 60 | ||
| XM_009298464.3 | XP_009296739.1 | 12 - 13 | 2737 | 60 | ||
| XM_009298468.3 | XP_009296743.1 | 14 - 15 | 2472 | 60 | ||
| XM_021481410.1 | XP_021337085.1 | 15 - 16 | 3104 | 60 | ||
| XM_021481419.1 | XP_021337094.1 | 13 - 14 | 2749 | 60 | ||
| XM_021481437.1 | XP_021337112.1 | 12 - 13 | 2448 | 60 | ||
| XM_021481438.1 | XP_021337113.1 | 11 - 12 | 2430 | 60 | ||
| XM_021481441.1 | XP_021337116.1 | 12 - 13 | 2448 | 60 | ||
| XM_021481443.1 | XP_021337118.1 | 11 - 12 | 2430 | 60 | ||
| GenBank mRNA | BC134804.1 | - | 12 - 13 | 2689 | 60 | |
Set Membership: |
|