Search
Search

clockb| Gene Symbol: clockb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178295.2 | NP_840080.2 | 7 - 8 | 726 | 94 | |
| XM_005168339.4 | XP_005168396.1 | 8 - 9 | 1110 | 94 | ||
| GenBank mRNA | AB087256.1 | - | 7 - 8 | 636 | 94 | |
| BC162682.1 | - | 7 - 8 | 716 | 94 | ||
Set Membership: |
|