Search
Search

hsd17b12b| Gene Symbol: hsd17b12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199613.1 | NP_955907.1 | 6 - 7 | 605 | 116 | |
| GenBank mRNA | AY551080.1 | - | 6 - 7 | 495 | 116 | |
| BC059617.1 | - | 6 - 7 | 605 | 116 | ||
| BC152213.1 | - | 6 - 7 | 614 | 116 | ||
| BC154318.1 | - | 6 - 7 | 577 | 116 | ||
Set Membership: |
|