Search
Search

rps6kal| Gene Symbol: rps6kal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200073.1 | NP_956367.1 | 17 - 18 | 1708 | 76 | |
| XM_009307718.2 | XP_009305993.1 | 12 - 13 | 1205 | 76 | ||
| XM_021480984.1 | XP_021336659.1 | 10 - 11 | 1013 | 76 | ||
| GenBank mRNA | BC045478.1 | - | 17 - 18 | 1813 | 76 | |
| BC057467.1 | - | 17 - 18 | 1708 | 76 | ||
Set Membership: |
|