Search
Search

nfe2l1a| Gene Symbol: nfe2l1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212855.2 | NP_998020.2 | 5 - 6 | 1112 | 98 | |
| XM_021469979.1 | XP_021325654.1 | 5 - 6 | 1171 | 98 | ||
| XM_021469986.1 | XP_021325661.1 | 5 - 6 | 1171 | 98 | ||
| GenBank mRNA | AY423002.1 | - | 5 - 6 | 1108 | 98 | |
| BC162497.1 | - | 5 - 6 | 1100 | 98 | ||
| BC162503.1 | - | 5 - 6 | 1100 | 98 | ||
| JX867114.1 | - | 4 - 5 | 919 | 98 | ||
Set Membership: |
|