Search
Search

tspan7| Gene Symbol: tspan7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214774.2 | NP_999939.1 | 6 - 7 | 849 | 86 | |
| GenBank mRNA | BC055255.1 | - | 6 - 7 | 789 | 86 | |
| BC097054.1 | - | 6 - 7 | 837 | 86 | ||
| BC165114.1 | - | 6 - 7 | 721 | 86 | ||
Set Membership: |
|