Search
Search

acaa1| Gene Symbol: acaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002207.1 | NP_001002207.1 | 7 - 8 | 616 | 89 | |
| GenBank mRNA | BC072706.1 | - | 7 - 8 | 616 | 89 | |
| BC165883.1 | - | 7 - 8 | 645 | 89 | ||
Set Membership: |
|